siRNA Details
Gene Symbol: Cpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012930.1 | NP_037062.1 | 4 | 1062 | ||
GenBank mRNA | J05470.1 | - | 4 | 1062 | ||
U88295.1 | - | 4 | 1000 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
