siRNA Details
Gene Symbol: Cyp1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012940.2 | NP_037072.1 | 3 | 1848 | ||
GenBank mRNA | U09540.1 | - | 1848 | |||
X78583.1 | - | 2 | 538 | |||
X83867.1 | - | 3 | 1849 |

More Information
Other Species Targeted

Gene Ontology Categories:
