siRNA Details
Gene Symbol: Hbegf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012945.1 | NP_037077.1 | 3 | 378 | ||
GenBank mRNA | L05489.1 | - | 3 | 378 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
