siRNA Details
Gene Symbol: Eef2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012947.2 | NP_037079.1 | 1 | 190 | ||
XM_006230156.2 | XP_006230218.1 | 2 | 514 | |||
XM_006230157.2 | XP_006230219.1 | 2 | 911 | |||
XM_006230158.2 | XP_006230220.1 | 2 | 1591 | |||
GenBank mRNA | BC061825.1 | - | 2 | 445 | 5599075 | |
U93849.1 | - | 1 | 190 | |||
X96426.1 | - | 480 |
