siRNA Details
Gene Symbol: Eef2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012947.2 | NP_037079.1 | 1 | 179 | ||
XM_006230156.2 | XP_006230218.1 | 2 | 503 | |||
XM_006230157.2 | XP_006230219.1 | 2 | 900 | |||
XM_006230158.2 | XP_006230220.1 | 2 | 1580 | |||
GenBank mRNA | BC061825.1 | - | 2 | 434 | 5599075 | |
U93849.1 | - | 1 | 179 | |||
X96426.1 | - | 469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
