siRNA Details
Gene Symbol: SLC6A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134367.3 | NP_001127839.2 | 12, 13 | 1771 | ||
NM_003043.5 | NP_003034.2 | 12, 13 | 1775 | |||
NR_103507.2 | 11, 12 | 1650 | ||||
XM_006713307.2 | XP_006713370.1 | 11, 12 | 1579 | |||
XM_011534030.2 | XP_011532332.1 | 11, 12 | 1532 | |||
XM_011534031.2 | XP_011532333.1 | 13, 14 | 1903 | |||
GenBank mRNA | AB209172.1 | - | 11, 12 | 1576 | ||
AK308417.1 | - | 10, 11 | 1251 | |||
BC137128.1 | - | 12, 13 | 1722 | 9021125 | ||
BC137129.1 | - | 12, 13 | 1718 | 9021126 | ||
U09220.1 | - | 10, 11 | 1450 | |||
U16120.1 | - | 13, 14 | 1826 | |||
Z18956.1 | - | 10, 11 | 1469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
