siRNA Details
Gene Symbol: Fshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199237.1 | NP_954707.1 | 7, 8 | 672 | ||
XM_008764476.1 | XP_008762698.1 | 5, 6 | 332 | |||
GenBank mRNA | L02842.1 | - | 7, 8 | 672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
