siRNA Details
Gene Symbol: Glrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053296.1 | NP_445748.1 | 3, 4 | 301 | ||
XM_006232506.2 | XP_006232568.1 | 4, 5 | 447 | |||
GenBank mRNA | AJ310839.1 | - | 3, 4 | 301 | ||
FQ212388.1 | - | 4, 5 | 452 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001281969, NM_010298, U09399, X81202, X81203, and others.
Gene Ontology Categories:
