siRNA Details
Gene Symbol: Icam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012967.1 | NP_037099.1 | 7 | 1609 | ||
GenBank mRNA | BC081837.1 | - | 7 | 1621 | 7122397 | |
D00913.1 | - | 7 | 1609 | |||
FQ228241.1 | - | 7 | 1629 |

More Information
Other Species Targeted

Gene Ontology Categories:
