siRNA Details
Gene Symbol: Lamb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012974.1 | NP_037106.1 | 7 | 751 | ||
XM_006243709.2 | XP_006243771.1 | 7 | 796 | |||
GenBank mRNA | BC088400.1 | - | 7 | 750 | 7107264 | |
X16563.1 | - | 7 | 751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
