siRNA Details
Gene Symbol: Myo1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173101.1 | NP_775124.1 | 21 | 2896 | ||
GenBank mRNA | X74815.1 | - | 21 | 2896 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_181072, AK166477, BC051391, BG076620, AK018649.
Gene Ontology Categories:
