siRNA Details
Gene Symbol: Myo1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173101.1 | NP_775124.1 | 4 | 543 | ||
GenBank mRNA | X74815.1 | - | 4 | 543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
