siRNA Details
Gene Symbol: Nes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012987.1 | NP_037119.1 | 5 | 5150 | ||
XM_006232600.2 | XP_006232662.1 | 4 | 5732 | |||
GenBank mRNA | AF538924.1 | - | 4 | 5287 | ||
M34384.1 | - | 5 | 5150 |
Gene Symbol: LOC100910255 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232557.2 | XP_006232619.1 | 4 | 5733 |
