siRNA Details
Gene Symbol: Nxph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012994.2 | NP_037126.1 | 3 | 1027 | ||
GenBank mRNA | BC105770.1 | - | 3 | 1027 | 7930348 | |
FQ213736.1 | - | 1 | 186 | |||
L27867.1 | - | 2 | 933 |
