siRNA Details
Gene Symbol: Nxph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012994.2 | NP_037126.1 | 3 | 867 | ||
GenBank mRNA | BC105770.1 | - | 3 | 867 | 7930348 | |
FQ213736.1 | - | 1 | 26 | |||
L27867.1 | - | 2 | 773 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008751, AK083014, AK047847, AK163438, BC079608, and others.
Gene Ontology Categories:
