siRNA Details
Gene Symbol: Pcsk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012999.1 | NP_037131.1 | 7 | 866 | ||
GenBank mRNA | D31854.1 | - | 5 | 487 | ||
L31894.1 | - | 7 | 866 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
