siRNA Details
Gene Symbol: Psap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190236.1 | NP_001177165.1 | 3 | 282 | ||
NM_001190237.1 | NP_001177166.1 | 3 | 282 | |||
NM_001190238.1 | NP_001177167.1 | 3 | 282 | |||
NM_013013.2 | NP_037145.2 | 3 | 282 | |||
XM_006256403.1 | XP_006256465.1 | 3 | 294 | |||
GenBank mRNA | BC061759.1 | - | 3 | 216 | 5600088 | |
CK365248.1 | - | 3 | 282 | |||
CO403947.1 | - | 1 | 50 | |||
FQ217536.1 | - | 3 | 233 | |||
M19936.1 | - | 3 | 232 | |||
S81353.1 | - | 3 | 232 |
