siRNA Details
Gene Symbol: Rab12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013017.1 | NP_037149.1 | 394 | |||
GenBank mRNA | M83676.1 | - | 3 | 189 |

More Information
Other Species Targeted

Gene Ontology Categories:
