siRNA Details
Gene Symbol: Rps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277243.1 | NP_001264172.1 | 5 | 587 | ||
GenBank mRNA | BC158718.1 | - | 5 | 545 | 9089281 | |
FM052146.1 | - | 5 | 543 | |||
FQ209785.1 | - | 5 | 561 | |||
FQ210357.1 | - | 5 | 561 | |||
FQ210491.1 | - | 5 | 544 | |||
FQ217060.1 | - | 5 | 561 | |||
FQ217574.1 | - | 5 | 560 | |||
FQ219286.1 | - | 5 | 561 | |||
FQ221071.1 | - | 5 | 561 | |||
FQ221219.1 | - | 5 | 561 | |||
FQ221255.1 | - | 5 | 561 | |||
FQ221284.1 | - | 5 | 561 | |||
FQ221608.1 | - | 5 | 561 | |||
FQ222025.1 | - | 5 | 561 | |||
FQ222269.1 | - | 5 | 561 | |||
FQ222365.1 | - | 5 | 561 | |||
FQ222894.1 | - | 5 | 561 | |||
FQ223194.1 | - | 5 | 606 | |||
FQ224029.1 | - | 5 | 561 | |||
FQ225463.1 | - | 5 | 561 | |||
FQ226199.1 | - | 5 | 561 | |||
FQ226257.1 | - | 5 | 564 | |||
FQ226702.1 | - | 5 | 581 | |||
FQ227598.1 | - | 5 | 561 | |||
FQ228474.1 | - | 5 | 561 | |||
FQ230232.1 | - | 5 | 560 | |||
FQ232912.1 | - | 5 | 561 | |||
X58465.1 | - | 5 | 561 |

More Information
Other Species Targeted

Gene Ontology Categories:
