siRNA Details
Gene Symbol: Sbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013024.2 | NP_037156.2 | 4 | 575 | ||
XM_008767547.1 | XP_008765769.1 | 3 | 561 | |||
XM_008767548.1 | XP_008765770.1 | 3 | 381 | |||
GenBank mRNA | CA339008.1 | - | 4 | 575 | ||
J02675.1 | - | 3 | 585 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
