siRNA Details
Gene Symbol: Ccl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013025.2 | NP_037157.2 | 2 | 212 | ||
GenBank mRNA | EF121994.1 | - | 2 | 143 | ||
EF121995.1 | - | 2 | 143 | |||
U06435.1 | - | 2 | 143 | |||
U22414.1 | - | 2 | 212 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
