siRNA Details
Gene Symbol: Slc34a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013030.1 | NP_037162.1 | 6 | 618 | ||
GenBank mRNA | AB013453.1 | - | 6 | 618 | ||
BC078876.1 | - | 6 | 612 | 7109613 | ||
L13257.1 | - | 6 | 618 |
