siRNA Details
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013034.4 | NP_037166.2 | 12 | 1799 | ||
XM_008767950.1 | XP_008766172.1 | 13 | 1855 | |||
GenBank mRNA | BF548147.1 | - | 2 | 126 | ||
M79450.1 | - | 14 | 1920 | |||
X63253.1 | - | 12 | 1807 | |||
X63995.1 | - | 13 | 1852 | |||
Y11024.1 | - | 12 | 1692 |
