siRNA Details
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013034.4 | NP_037166.2 | 1 | 431 | ||
XM_008767950.1 | XP_008766172.1 | 2 | 487 | |||
GenBank mRNA | CB736426.1 | - | 1 | 412 | ||
M79450.1 | - | 3 | 555 | |||
X63253.1 | - | 1 | 439 | |||
X63995.1 | - | 2 | 484 | |||
Y11024.1 | - | 1 | 324 |
