siRNA Details
Gene Symbol: Abcc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013040.2 | NP_037172.2 | 15, 16 | 2175 | ||
GenBank mRNA | AB045281.1 | - | 14, 15 | 1963 | ||
AF019628.1 | - | 15, 16 | 2175 | |||
AF087838.1 | - | 15, 16 | 2175 | |||
D83598.1 | - | 15, 16 | 2175 |
