siRNA Details
Gene Symbol: Tmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013044.2 | NP_037176.2 | 8 | 834 | ||
GenBank mRNA | BC072521.1 | - | 8 | 834 | 7105834 | |
U59241.1 | - | 7 | 790 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013044.2 | NP_037176.2 | 8 | 834 | ||
GenBank mRNA | BC072521.1 | - | 8 | 834 | 7105834 | |
U59241.1 | - | 7 | 790 |