siRNA Details
Gene Symbol: Ube2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013050.1 | NP_037182.1 | 5, 6 | 406 | ||
XM_006245918.2 | XP_006245980.1 | 5, 6 | 406 | |||
XM_006245919.2 | XP_006245981.1 | 5, 6 | 1086 | |||
XM_006245920.2 | XP_006245982.1 | 5, 6 | 481 | |||
XM_008767549.1 | XP_008765771.1 | 5, 6 | 538 | |||
GenBank mRNA | BC086324.1 | - | 5, 6 | 404 | 7305067 | |
BC086592.1 | - | 5, 6 | 404 | 7310403 | ||
FQ226760.1 | - | 5, 6 | 481 | |||
FQ228426.1 | - | 5, 6 | 391 | |||
FQ229169.1 | - | 5, 6 | 391 | |||
U54632.1 | - | 5, 6 | 406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
