siRNA Details
Gene Symbol: Ubtf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105723.3 | NP_001099193.2 | 14 | 1653 | ||
NM_001127690.2 | NP_001121162.1 | 15 | 1764 | |||
XM_006247262.2 | XP_006247324.1 | 15 | 1761 | |||
XM_006247263.2 | XP_006247325.1 | 15 | 2012 | |||
XM_006247264.2 | XP_006247326.1 | 15 | 2582 | |||
XM_008767951.1 | XP_008766173.1 | 14 | 1650 | |||
XM_008767952.1 | XP_008766174.1 | 13 | 1888 | |||
GenBank mRNA | FQ224307.1 | - | 15 | 1675 | ||
M61725.1 | - | 13 | 1709 | |||
M61726.1 | - | 14 | 1820 |
