siRNA Details
Gene Symbol: Scgb1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013051.1 | NP_037183.1 | 2 | 273 | ||
GenBank mRNA | BC069174.1 | - | 2 | 263 | 7096714 | |
J05536.1 | - | 2 | 273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
