siRNA Details
Gene Symbol: Id2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013060.3 | NP_037192.1 | 1 | 192 | ||
GenBank mRNA | AY321351.1 | - | 325 | |||
BC086391.1 | - | 187 | 7303450 | |||
D10863.1 | - | 1 | 192 | |||
FQ212969.1 | - | 210 | ||||
FQ213471.1 | - | 210 | ||||
FQ213662.1 | - | 210 | ||||
FQ220574.1 | - | 1 | 210 | |||
FQ227397.1 | - | 1 | 210 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010496, M69293, BC053699, AK135547, AK146346, and others.
Gene Ontology Categories:
