siRNA Details
Gene Symbol: Hcrtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013064.1 | NP_037196.1 | 3 | 683 | ||
GenBank mRNA | AF041244.1 | - | 3 | 683 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hcrtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013064.1 | NP_037196.1 | 3 | 683 | ||
GenBank mRNA | AF041244.1 | - | 3 | 683 |
Set Membership: |
![]() |