siRNA Details
Gene Symbol: Rpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB100593.1 | - | 7 | 1222 | ||
AB100594.1 | - | 7 | 1222 | |||
BC061756.1 | - | 7 | 1226 | 5600029 | ||
FQ229660.1 | - | 1 | 30 | |||
X05300.1 | - | 7 | 1227 |
