siRNA Details
Gene Symbol: Rasa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105724.1 | NP_001099194.1 | 23 | 2354 | ||
GenBank mRNA | D30734.1 | - | 23 | 2398 |
Gene Symbol: LOC103693168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766600.1 | XP_008764822.1 | 1 | 70 |
