siRNA Details
Gene Symbol: Utrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013070.1 | NP_037202.1 | 17 | 2146 | ||
XM_006227637.2 | XP_006227699.1 | 16 | 2321 | |||
XM_008758593.1 | XP_008756815.1 | 17 | 2245 | |||
XM_008758594.1 | XP_008756816.1 | 17 | 2142 | |||
XM_008758595.1 | XP_008756817.1 | 16 | 2321 | |||
XM_008758596.1 | XP_008756818.1 | 16 | 2314 | |||
XM_008758597.1 | XP_008756819.1 | 16 | 2320 | |||
GenBank mRNA | AJ002967.1 | - | 17 | 2146 |
