siRNA Details
Gene Symbol: Hoxa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013075.1 | NP_037207.1 | 2 | 922 | ||
XM_008773708.1 | XP_008771930.1 | 3 | 838 | |||
GenBank mRNA | U93092.1 | - | 2 | 922 |
Gene Symbol: NEWGENE_2812 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762937.1 | XP_008761159.1 | 2 | 1041 | ||
XM_008762938.1 | XP_008761160.1 | 3 | 838 |
