siRNA Details
Gene Symbol: Tub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013077.1 | NP_037209.1 | 8 | 935 | ||
XM_008759690.1 | XP_008757912.1 | 8 | 1211 | |||
XM_008759691.1 | XP_008757913.1 | 7 | 1044 | |||
GenBank mRNA | AB011544.1 | - | 8 | 935 | ||
U92546.1 | - | 1 | 64 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_021885, U54643, AK147700, U52824, BC096554, and others.
Gene Ontology Categories:
