siRNA Details
Gene Symbol: Otc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013078.1 | NP_037210.1 | 8 | 829 | ||
GenBank mRNA | BC088158.1 | - | 8 | 795 | 7325875 | |
FQ211098.1 | - | 8 | 813 | |||
FQ219657.1 | - | 8 | 792 | |||
K03040.1 | - | 8 | 787 | |||
M11266.1 | - | 8 | 829 |
