siRNA Details
Gene Symbol: Sdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013082.3 | NP_037214.1 | 2, 3 | 562 | ||
XM_006241518.2 | XP_006241580.1 | 2, 3 | 774 | |||
GenBank mRNA | BC070890.1 | - | 2, 3 | 352 | 7098631 | |
FQ218442.1 | - | 2, 3 | 295 | |||
FQ219283.1 | - | 2, 3 | 596 | |||
FQ219510.1 | - | 2, 3 | 594 | |||
FQ222974.1 | - | 2, 3 | 644 | |||
M81687.1 | - | 2, 3 | 562 |
