siRNA Details
Gene Symbol: Vamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013090.2 | NP_037222.2 | 3 | 404 | ||
GenBank mRNA | AF498262.1 | - | 2 | 270 | ||
BC092206.1 | - | 3 | 302 | 7313343 | ||
CB582258.1 | - | 3 | 404 | |||
EF653274.1 | - | 3 | 329 | |||
EF653275.1 | - | 3 | 329 | |||
M24104.1 | - | 3 | 367 | |||
U74621.1 | - | 3 | 302 |
