siRNA Details
Gene Symbol: Pkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053297.2 | NP_445749.1 | 9 | 1341 | ||
XM_006243187.2 | XP_006243249.1 | 9 | 1571 | |||
XM_006243190.2 | XP_006243252.1 | 10 | 1643 | |||
GenBank mRNA | FQ223721.1 | - | 9 | 1349 | ||
FQ224316.1 | - | 6 | 999 | |||
X15800.1 | - | 9 | 1343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
