siRNA Details
Gene Symbol: Prkaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100922.1 | NP_001094392.1 | 3 | 336 | ||
XM_006255219.1 | XP_006255281.1 | 3 | 270 | |||
GenBank mRNA | AY375243.1 | - | 3 | 182 | ||
BC129128.1 | - | 3 | 336 | 7314651 | ||
X57986.1 | - | 3 | 246 |
