siRNA Details
Gene Symbol: Ptger1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278475.1 | NP_001265404.1 | 3 | 1435 | ||
XM_008772362.1 | XP_008770584.1 | 3 | 2087 | |||
GenBank mRNA | D88751.1 | - | 2 | 1727 | ||
D88752.1 | - | 1 | 2152 | |||
U68037.1 | - | 2 | 1143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
