siRNA Details
Gene Symbol: Gnai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013106.1 | NP_037238.1 | 4 | 514 | ||
GenBank mRNA | DQ120473.1 | - | 4 | 407 | ||
DQ120474.1 | - | 4 | 407 | |||
FQ222046.1 | - | 4 | 577 | |||
J03219.1 | - | 4 | 523 | |||
M20713.1 | - | 4 | 514 |
