siRNA Details
Gene Symbol: Gja4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021654.2 | NP_067686.1 | 2 | 392 | ||
GenBank mRNA | BC078837.1 | - | 2 | 392 | 7128770 | |
BC086576.1 | - | 1 | 323 | 7306623 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gja4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021654.2 | NP_067686.1 | 2 | 392 | ||
GenBank mRNA | BC078837.1 | - | 2 | 392 | 7128770 | |
BC086576.1 | - | 1 | 323 | 7306623 |