siRNA Details
Gene Symbol: Gckr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013120.2 | NP_037252.1 | 16 | 1583 | ||
GenBank mRNA | BC081843.1 | - | 16 | 1583 | 7096237 | |
KJ026952.1 | - | 16 | 1406 | |||
X68497.1 | - | 16 | 1427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
