siRNA Details
Gene Symbol: Cd28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013121.1 | NP_037253.1 | 2 | 426 | ||
XM_008767078.1 | XP_008765300.1 | 2 | 1070 | |||
XM_008767079.1 | XP_008765301.1 | 3 | 513 | |||
GenBank mRNA | X55288.1 | - | 2 | 426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
