siRNA Details
Gene Symbol: Fn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019143.2 | NP_062016.2 | 26, 27 | 4452 | ||
XM_006245150.2 | XP_006245212.1 | 26, 27 | 4454 | |||
XM_006245151.2 | XP_006245213.1 | 26, 27 | 4453 | |||
XM_006245152.2 | XP_006245214.1 | 25, 26 | 4180 | |||
XM_006245153.2 | XP_006245215.1 | 25, 26 | 4180 | |||
XM_006245154.2 | XP_006245216.1 | 26, 27 | 4454 | |||
XM_006245155.2 | XP_006245217.1 | 23, 24 | 3826 | |||
XM_006245156.2 | XP_006245218.1 | 25, 26 | 4180 | |||
XM_006245157.2 | XP_006245219.1 | 25, 26 | 4180 | |||
XM_006245158.2 | XP_006245220.1 | 25, 26 | 4180 | |||
XM_006245159.2 | XP_006245221.1 | 25, 26 | 4180 | |||
GenBank mRNA | X15906.1 | - | 26, 27 | 4450 |
