siRNA Details
Gene Symbol: Pparg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145366.1 | NP_001138838.1 | 7 | 950 | ||
NM_001145367.1 | NP_001138839.1 | 7 | 1128 | |||
NM_013124.3 | NP_037256.1 | 6 | 925 | |||
XM_006237009.2 | XP_006237071.1 | 7 | 921 | |||
GenBank mRNA | AB011365.1 | - | 5 | 758 | ||
AB019561.1 | - | 6 | 839 | |||
AF156665.1 | - | 7 | 950 | |||
AF156666.1 | - | 6 | 887 | |||
AF246457.1 | - | 7 | 878 | |||
AF246458.1 | - | 7 | 918 | |||
Y12882.2 | - | 6 | 888 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
