siRNA Details
Gene Symbol: Pparg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145366.1 | NP_001138838.1 | 5 | 616 | ||
NM_001145367.1 | NP_001138839.1 | 5 | 794 | |||
NM_013124.3 | NP_037256.1 | 4 | 591 | |||
XM_006237009.2 | XP_006237071.1 | 5 | 587 | |||
GenBank mRNA | AB011365.1 | - | 3 | 424 | ||
AB019561.1 | - | 4 | 505 | |||
AF156665.1 | - | 5 | 616 | |||
AF156666.1 | - | 4 | 553 | |||
AF246457.1 | - | 5 | 544 | |||
AF246458.1 | - | 5 | 584 | |||
Y12882.2 | - | 4 | 554 |

More Information
Other Species Targeted

Gene Ontology Categories:
