siRNA Details
Gene Symbol: Glra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013133.1 | NP_037265.1 | 4 | 605 | ||
XM_006246378.2 | XP_006246440.1 | 4 | 757 | |||
GenBank mRNA | AJ310834.1 | - | 4 | 421 | ||
AJ310835.1 | - | 4 | 421 | |||
AJ310836.1 | - | 3 | 293 | |||
AY827463.1 | - | 4 | 421 | |||
D00833.1 | - | 4 | 605 | |||
X55246.1 | - | 4 | 509 | |||
Y00276.1 | - | 3 | 357 |
